Palladium, black ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

00659.03 00659.06
00659.03EA 382 EUR
00659.03 00659.06
Palladium, black ≥99.9% (metals basis)
Palladium

Purity: 99.9% CAS No.: 7440-05-3 Physical Form: S.A. typically 20m2/g Odour: Odourless UN No.: 3089 MDL No.: MFCD00011167

Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR