Magnesium ionophore VII, Selectophore™, function tested ionophore, Supelco®

Supplier: Merck

Synonyms: 4,13-[Bis(N-adamantylcarbamoyl)acetyl]-1,7,10,16,tetraoxa-4,13-diazacyclooctadecane, K22B5

M-Clarity™ quality level = MQ100

00744-10MG-F
00744-10MG-FEA 688 EUR
00744-10MG-F
Magnesium ionophore VII, Selectophore™, function tested ionophore, Supelco®
Magnesium ionophore VII
Formula: C₃₈H₆₀N₄O₈
Storage Temperature: Freezer
MDL Number: MFCD07785359
CAS Number: 156210-12-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae0c9dca10d45beced917c02ce2903582be73e8c&vid=030dcc9f0e1ec2d5acd36cea9c9a952219717528&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR