Bromotrispyrrolidinophosphonium hexafluorophosphate

Supplier: Novabiochem (Part of Merck)

Synonyms: Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, PyBrOP , Bromotripyrrolidinylphosphonium hexafluorophosphate, Bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)

8.51010.0100 8.51010.0025
01-62-0017-100EA 1290 EUR
01-62-0017-100 01-62-0017-25
Bromotrispyrrolidinophosphonium hexafluorophosphate
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate
Formula: C₁₂H₂₄BrF₆N₃P₂
MDL Number: MFCD00077412
CAS Number: 132705-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR