Fmoc-NH-(PEG)₂-COOH (20 atoms) 92 HPLC_ASSAY_METHOD, Novabiochem®

Supplier: Novabiochem (Part of Merck)

8.51031.0001 8.51031.8500 8.51031.0005
01-63-0141-1EA 699 EUR
01-63-0141-1 8.51031.8500 NOVA8.51031.0005
Fmoc-NH-(PEG)₂-COOH (20 atoms) 92 HPLC_ASSAY_METHOD, Novabiochem®
Fmoc-NH-(PEG)₂-COOH (20 atoms)

It is a bifunctional 20 atom PEG spacer. It can be introduced using standard coupling methods, such as PyBOP®, HBTU or TBTU and is compatible with standard TFA cleavage protocols.

Formula: C₂₉H₃₈N₂O₉
MW: 558.62 g/mol
MDL Number: MFCD17019373
CAS Number: 916585-44-9

Order Now

Specification Test Results

Colour White to slight yellow to beige
Appearance of substance Powder
Identity (IR) Passes test
Purity (TLC(0110)) ≥ 92%
Assay (HPLC, area %) ≥ 92.0% (a/a)
Storage Store at 2...8 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR