Chromatography Tubing, Stainless Steel
Supplier: Avantor
HICH01-SS-07-3MEA 118 EUR
HICH01-SS-07-3M HICH01-SS-20-3M 01-SS-05-3M 01-SS-30-5M 01-SS-10-3M 01-SS-40-3M 01-SS-07-10M 01-SS-10-10M 01-SS-20-10M 01-SS-30-10M 01-SS-40-10M
Chromatography Tubing, Stainless Steel
Tubing Chromatography Tubing
316 Stainless steel, precision bore ¹/₁₆" type tubing.
  • Operating temperature - no limit
  • May corrode with chloride salt or other buffers
  • May adsorb proteins
0,005 and 0,007" Int.Ø - suitable for minimal dead volume injector-column or column-detector connections of microbore or short columns containing 3 µm material. Sample and material must be filtered to avoid blockage.
0,10' Int.Ø - suitable for connection of analytical and semi-preparative columns containing 5 and 10 µm material. It should be used in all parts of the system where dead volumeMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable