Vibratory disc mill, RS 200
Supplier: Retsch GmbH
01.462.0171 01.462.0178 01.462.0179 01.462.0170 01.462.0192 01.462.0193 01.462.0263 01.462.0266 01.462.0002 20.730.0001 01.462.0353 01.462.0354 01.462.0355
01.462.0171EA 2710 EUR
01.462.0171 01.462.0178 01.462.0179 01.462.0170 01.462.0192 01.462.0193 412-2920 RETS01.462.0266 RETS01.462.0002 412-0258 412-0255 412-0256 412-0257
Vibratory disc mill, RS 200
Mills
The Vibratory Disc Mill RS200 is suitable for the extremely quick, loss-free and reproducible grinding of medium-hard, brittle and fibrous materials to analytical fineness. The RS200 is primarily used for sample preparation for spectral analysis.
  • Powerful stabilised-plane-drive
  • Analytical fineness in seconds with excellent reproducibility
  • One-button operation with graphics display
  • 10 parameter combinations as SOPs can be stored
  • Grinding tools in 4 different materials
  • IP 40
* depending on feed material and configuration/settiings