Ammonium acetate, Ultrapure

Supplier: VWR Chemicals
Warning

Synonyms: Acetic acid ammonium salt

0103-2.5KGEA 130 EUR
0103-2.5KG 0103-500G
Ammonium acetate, Ultrapure
Ammonium acetate
Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@280 nm (1 M, Water) 0.01%
Abs.@354 nm (1 M, Water) 0.01%
Chloride <0.0005%
Heavy Metals (as Pb) 0.005%
Insolubles <0.0005%
Iron <0.001%
Nitrate <0.001%
pH (5%, Water) @25 °C 6.7 - 7.3
Purity 97.0%
Residue after Ignition 0.01%
Sulphate <0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR