Polyethylene glycol 8,000 for biotechnology

Supplier: VWR Chemicals
Warning

0159-1KGEA 282 EUR
0159-1KG 0159-2.5KG 0159-500G
Polyethylene glycol 8,000 for biotechnology
Polyethylene glycol 8,000

Polyethylene glycol is used for the isolation of plasmid DNA and the precipitation of phage.

Formula: H(OCH₂CH₂)nOH
Boiling Pt: >250 °C
Density: 1,21 g/cm³ (20 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD01779615
CAS Number: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23e4f2ac7d7b08ad5e6814488bdedf96bd458a77&vid=93a10b0bbb73df577d5985efd8329219e5fe1647&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNase None Detected
pH (5%, Water) @25 °C 4.5 – 7.5
RNase None Detected
Solidification Point 55 – 64 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR