Ammonium sulphate ≥99.5%, Ultra Pure Grade

Supplier: VWR Chemicals

0191-1KGEA 178 EUR
0191-1KG 0191-10KG 0191-5KG 0191-12KG 0191-50KG
Ammonium sulphate ≥99.5%, Ultra Pure Grade
Ammonium sulphate

May be used for the precipitation or fractionation of proteins or for purification of antibodies. Useful for crystallographic analysis of nucleic acids and proteins.

Formula: (NH₄)₂SO₄
Melting Pt: 280 °C
Density: 1,78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Arsenic <0.0001%
Chloride ≤0.0005%
DNase none detected
Heavy Metals (as Pb) ≤0.0002%
Insolubles ≤0.005%
Iron ≤0.0005%
Magnesium Test Pass
pH (5%, Water) @25 °C 5 – 6
Phosphate ≤0.0005%
Protease none detected
Purity (Dry Basis) ≥99.5%
Residue on Ignition ≤0.005%
RNase none detected
Selenium ≤0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR