Bis(2-ethylhexyl)adipate, Supelco®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

02138-1ML 02138-25ML 02138-5ML
02138-1MLEA 35.2 EUR
02138-1ML 02138-25ML 02138-5ML
Bis(2-ethylhexyl)adipate, Supelco®
Bis(2-ethylhexyl) adipate
Formula: C₂₂H₄₂O₄
MDL Number: MFCD00009496
CAS Number: 103-23-1
EINECS: 203-090-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20b7b39899cb11df8b3142a6afff257d86d814f7&vid=a9e92c4e46be7355c19e4c6335d9856775dbb326&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR