Potassium hydroxide, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Potash caustic, Caustic potash

0222.1000EA 71 EUR
0222.1000
Potassium hydroxide, BAKER ANALYZED® ACS, J.T.Baker®
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay min. 86.0 %
Calcium (Ca) max. 0.005 %
Chloride (Cl) max. 0.001 %
Magnesium (Mg) max. 0.002 %
Sodium (Na) max. 0.05 %
Heavy Metals (as Ag) max. 0.001 %
Insoluble Matter max. 30 ppm
Potassium Carbonate (K2CO3) max. 0.5 %
Trace Impurities (in ppm)
Iron (Fe) max. 3
Mercury (Hg) max. 0.1
Nickel (Ni) max. 2
Nitrogen Compounds (as N) max. 3
Phosphate (PO4) max. 2
Sulfate (SO4) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR