Ethanol absolute (untaxed) ≥99.8%, Puriss. p.a. ACS, A15, without additive, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

02860-1L 02860-2.5L 02860-25L 02860-4X2.5L 02860-6X1L
02860-1LEA 72.7 EUR
02860-1L 02860-2.5L 02860-25L 02860-4X2.5L 02860-6X1L
Ethanol absolute (untaxed) ≥99.8%, Puriss. p.a. ACS, A15, without additive, Riedel-de Haën™
Ethanol
Formula: C₂H₅OH
Boiling Pt: 78…79 °C (1,013 hPa)
Melting Pt: –115 °C
Density: 0,790…0.791 g/cm3 (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Refractive index (n 20/D) 1.361 - 1.363
Density (D 20/4) 0.789 - 0.791
Water (Karl Fischer) Max. 0.2 %
Non-volatile matter Max. 0.001 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.1 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 1 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Acetone (GC) Max. 10 ppm
Acetone and 2-propanol according to ACS complying
Aldehydes (as CH₃CHO) Max. 10 ppm
Formaldehyde Max. 10 ppm
APHA Max. 10
Refractive Index n20/D 1.3600 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR