Copper(II) sulphate pentahydrate 98-102 ACS

Supplier: VWR Chemicals
Warning

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

0330-2.5KGEA 276 EUR
0330-2.5KG 0330-500G
Copper(II) sulphate pentahydrate 98-102 ACS
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9 III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Calcium <0.005%
Chloride ≤0.001%
Insolubles ≤0.005%
Iron ≤0.003%
Nickel ≤0.005%
Nitrogen Compounds (as N) ≤0.002%
Potassium ≤0.01%
Purity 98% – 102%
Sodium ≤0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR