N(α)-Boc-L-tryptophan

Supplier: Novabiochem (Part of Merck)

Synonyms: Boc-Trp-OH , Boc-Trp-OH

8.53038.0100 8.53038.0025
04-12-0006-100EA 333 EUR
04-12-0006-100 04-12-0006-25
N(α)-Boc-L-tryptophan
N(α)-Boc-L-tryptophan
Formula: C₁₆H₂₀N₂O₄
MW: 304.3 g/mol
MDL Number: MFCD00065595
CAS Number: 13139-14-5
EINECS: 236-072-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR