Sodium hydroxide, BAKER, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Caustic soda

0404.1000EA 38.4 EUR
0404.1000
Sodium hydroxide, BAKER, Laboratory Reagent, J.T.Baker®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8 II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Assay min. 97 %
Carbonate (as Na2CO3) max. 1 %
Aluminium (Al) max. 0.002 %
Chloride (Cl) max. 0.01 %
Iron (Fe) max. 0.002 %
Heavy Metals (as Pb) max. 0.002 %
Sulfate (SO4) max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR