Organic reference standard, Acesulfame K, for food analysis, ≥99,0%, Supelco®
Supplier: Merck
04054-25G 04054-100G
04054-25GEA 90.3 EUR
04054-25G 04054-100G
Organic reference standard, Acesulfame K, for food analysis, ≥99,0%, Supelco®
Standards Organic Standards
Heat-stable sweetener that has not been suspected to cause cancer nor be genotoxic. Allelic variation of the Tas1r3 gene affects behavioral taste responses to this molecule, suggesting that it is a T1R3 receptor ligand.