Diammonium hydrogen orthophosphate 99-102%, Puriss., meets analytical specification of NF, Fluka™

Supplier: Honeywell Chemicals

04211-1KG 04211-25KG
04211-1KGEA 78.4 EUR
04211-1KG 04211-25KG
Diammonium hydrogen orthophosphate 99-102%, Puriss., meets analytical specification of NF, Fluka™
di-Ammonium hydrogen orthophosphate
Formula: (NH₄)₂HPO₄
Melting Pt: 155 °C
Density: 1,62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
pH (1 %,25°C) 7.6 - 8.2
Arsenic (As) Max. 2 ppm
Iron (Fe) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.002 %
Sulfate (SO₄) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR