Phenol, BAKER, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

0505.1000EA 132.79 EUR
0505.1000
Phenol, BAKER, J.T.Baker®
Phenol
Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.7
Melting Pt: 40.8
Density: 1.0722 g/cm³ (20 °C)
Flash Pt: 81
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
REACH: 01-2119471329-32
Merck Index: 12,07390

Order Now

Specification Test Results

Meets Reagent Specifications for testing Ph. Eur. monographs.
Assay 99.0 - 100.5 %
Acidity passes test
Appearance of solution passes test
Freezing Point min. 39.5 °C
Identification passes test
Residue on Evaporation max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR