Guanidinium hydrochloride 99.5-101.0%, NF-GenAR® for biotechnology, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

0506-06EA 0 EUR
0506-06
Guanidinium hydrochloride 99.5-101.0%, NF-GenAR® for biotechnology, Macron Fine Chemicals™
Guanidinium hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product ≤0.3%
CAUTION: For Manufacturing, processing or repackaging 99.5 - 101.0%
Appearance Passes Test
Water-Insoluble Matter ≤0.3 %
Assay (NH₂C(:NH)NH₂·HCl) 99.5 - 101.0 %
Ultraviolet Absorbance - 275 nm, For Information Only
Ultraviolet Absorbance - 260 nm, For Information Only
Loss on Drying ≤0.5 %
Trace Impurities - Iron (Fe) ≤2.0 ppm
MANUFACTURED EXCLUSIVELY FOR AVANTOR PERFORMANCE MATERIALS, LLC., BY
BIOSPECTRA, INC., STROUDSBURG, PA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR