Ammonium hydrogen difluoride ≥96.0%, flakes, BAKER ANALYZED®, J.T.Baker®

Supplier: Avantor
Danger


Management of Change (MOC) category = N

0619-05EA 245.41 EUR
0619-05 0619-07 0619-09
Ammonium hydrogen difluoride ≥96.0%, flakes, BAKER ANALYZED®, J.T.Baker®
Ammonium hydrogen difluoride
Formula: NH₄HF₂
Melting Pt: 124,6 °C
Density: 0,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NH₄FHF) ≥96.0 %
Insoluble Matter ≤ 0.02 %
pH of 5% Solution at 25°C 4.0 - 6.5
Heavy Metals (as Pb) ≤ 0.002 %
Moisture (as H₂O) ≤ 0.4 %
Product may contain black specks that are indigenous to product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR