N-Lauroylsarcosine sodium salt, Reagent Grade

Supplier: VWR Chemicals
Danger

0719-1KGEA 396 EUR
0719-1KG 0719-500G
N-Lauroylsarcosine sodium salt, Reagent Grade
N-Lauroylsarcosine sodium salt

Anionic - negatively-charged (carboxylates, sulfates or sulfonates in alkali metal form). Utilised for applications ranging from solubilising membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.


  • Very fine white crystalline powder

Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Melting Pt: 45…47 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
EINECS: 205-281-5
UN: 2811
ADR: 6.1,II
Merck Index: 12,04379
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0126d80345cce5dd675a83796c379bd44cebacef&vid=48c74917d6a93a32467317ef6c6fdbf24f3b6dc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

pH (10%, Water) @25 °C 7.5 – 8.5
Purity >92.0 %
Sodium Laurate ≤ 4.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR