4-Amino-N,N-dimethylaniline, Supelco®

Supplier: Merck
Danger

Synonyms: N,N-Dimethylbenzene-p-diamine, N,N-Dimethyl-1,4-phenylenediamine, N,N-Dimethylbenzene-1,4-diamine, N,N-Dimethyl-p-phenylenediamine

M-Clarity™ quality level = MQ100

07750-25G
07750-25GEA 48.5 EUR
07750-25G
4-Amino-N,N-dimethylaniline, Supelco®
4-Amino-N,N-dimethylaniline
Formula: C₈H₁₂N₂
MW: 136.2 g/mol
Boiling Pt: 262
Melting Pt: 35
Density: 1.09 g/cm³ (20 °C)
Flash Pt: 130
Storage Temperature: Ambient
MDL Number: MFCD00007860
CAS Number: 99-98-9
EINECS: 202-807-5
UN: 2811
ADR: 6.1 II
Merck Index: 13,33280
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=512f18d480fb3ad9b97af100d9f182d7f3497da9&vid=143a7125eec59aa2172fc921c49fc3514e396c43&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR