(±)-2-Ethyl-1-hexanol, Supelco®

Supplier: Merck
Warning

Synonyms: 2-Ethylhexan-1-ol

M-Clarity™ quality level = MQ100

08607-1ML-F 08607-5ML-F
08607-1ML-FEA 27.6 EUR
08607-1ML-F 08607-5ML-F
(±)-2-Ethyl-1-hexanol, Supelco®
(±)-2-Ethyl-1-hexanol
Formula: CH₃(CH₂)₃CH(C₂H₅)CH₂OH
MW: 130.23 g/mol
Boiling Pt: 183
Melting Pt: –76 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 81
Storage Temperature: Ambient
MDL Number: MFCD00004746
CAS Number: 104-76-7
EINECS: 203-234-3
REACH: 01-2119487289-20
Merck Index: 13,03842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60d84ececfb8331f731326b4f45a5aff19419311&vid=b147be53f238c69bee41eab5fd907f2a5ec65ba3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR