Ammonium carbamate, Supelco®

Supplier: Merck
Warning

Synonyms: Carbamic acid ammonium salt

M-Clarity™ quality level = MQ100

09699-1KG
09699-1KGEA 337 EUR
09699-1KG
Ammonium carbamate, Supelco®
Ammonium carbamate
Formula: H₂NCO₂NH₄
MDL Number: MFCD00013010
CAS Number: 1111-78-0
EINECS: 214-185-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9659246237fa206751b5ac3e9224119200d3c20a&vid=14783bdc7b4b40324e328367a26c2d311f0f794c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR