Formic acid : Triethylamine concentrate (1:1) 2 M:2 M in water for HPLC, Fluka™

Supplier: Honeywell Chemicals
Danger

09752-100ML
09752-100MLEA 360 EUR
09752-100ML
Formic acid : Triethylamine concentrate (1:1) 2 M:2 M in water for HPLC, Fluka™
Formic acid : Triethylamine
Boiling Pt: 89 °C
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134943
UN: 1760
ADR: 8 II

Order Now

Specification Test Results

Density (D 20/4) 1.005 - 1.014
Refractive index (n 20/D) 1.354 - 1.374
Suitability test (HPLC) complying
Absorbance 500 nm Max. 0.01
Absorbance at 290 nm Max. 0.02
Absorbance 280 nm Max. 0.03
Absorbance at 270 nm Max. 0.03
Absorbance at 260 nm Max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR