Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

1.00014.1000 1.00014.1011 1.00014.2500 1.00014.2511 1.00014.5000 1.00014.6010 1.00014.6025 1.00014.6190 1.00014.9180 1.00014.9900 1.00014.9912 1.00014.4000
1.00014.1000EA 31.14 EUR
1.00014.1000 1.00014.1011 1.00014.2500 1.00014.2511 1.00014.5000 1.00014.6010 1.00014.6025 1.00014.6190 1.00014.9180 1.00014.9900 1.00014.9912 1.00014.4000
Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Acetone

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. Merck declares their EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph. Eur.) and also with the ISO standards.

Formula: CH₃COCH₃
Boiling Pt: 56,2 °C (1013 hPa)
Melting Pt: –95,4 °C
Density: 0,792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -95.4 °C
Molar Mass 58.08 g/mol
Boiling Point 56.2 °C (1013 hPa)
Vapor Pressure 233 hPa (20 °C)
Refractive Index 1.35868 (20 °C)
Explosion Limit 2.6 - 12.8%(V)
Density 0.79 g/cm³ (20 °C)
pH 5 - 6 (395 g/L, H₂O, 20 °C)
Ignition Point 465 °C DIN 51794
Water Absorption 1000 g/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR