Acetone, Uvasol® Spectroscopy, Supelco®

Supplier: Merck
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

MQ100

1.00022.0500 1.00022.2500
1.00022.0500EA 84.2 EUR
1.00022.0500 1.00022.2500
Acetone, Uvasol® Spectroscopy, Supelco®
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 C) soluble
Melting Point -95.4 C
Molar Mass 58.08
Boiling Point 56.2 C
Vapor Pressure 233
Flash Point -18 C
Refractive Index 1.35686
Explosion Limit 2.6 - 12.8 %
Density 0.79 g/cm3
pH 5 - 6
Ignition Point 465 C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR