Citric acid monohydrate 99,5-100,5 alkalimetry on anhydrous substance, powder, EMPROVE® ESSENTIAL USP, JP, E330, Ph. Eur., BP, FCC suitable for use as excipient, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

1.00243.1000 1.00243.9029 1.00243.9025 1.00243.9026
1.00243.1000EA 324 EUR
1.00243.1000 1.00243.9029 1.00243.9025 1.00243.9026
Citric acid monohydrate 99,5-100,5 alkalimetry on anhydrous substance, powder, EMPROVE® ESSENTIAL USP, JP, E330, Ph. Eur., BP, FCC suitable for use as excipient, SAFC®
Citric acid monohydrate
Formula: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Melting Pt: 135…152 °C
Density: 1.552 g/cm³ (20 °C)
Flash Pt: 173.9 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (alkalimetric, calculated on anhydrous substance) 99.5 - 100.5 %
Identity (IR-spectrum) passes test
Appearance white to almost white, fine powder
Appearance of solution (200 g/l; water) clear (≤ 3 NTU) and not more intense in color than reference solution Y₇, BY₇ or GY₇
Sulfate (SO₄) ≤ 50 ppm
Heavy metals (as Pb) ≤ 5 ppm
Al (Aluminium) ≤ 0.2 ppm
As (Arsenic) ≤ 1 ppm
Hg (Mercury) ≤ 1 ppm
Pb (Lead) ≤ 0.5 ppm
Oxalates (as C₂H₂O₄) ≤ 100 ppm
Residual solvents (ICH (Q3C)) excluded by manufacturing process
Readily carbonisable substance passes test
Sulfated ash (600 °C) ≤ 0.05 %
Water (according to Karl Fischer) 7.5 - 8.8 %
Particle size (> 0.3 mm) ≤ 5.0 %
Bacterial endotoxins < 0.5
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph. Eur., BP, JP, USP, E 330, FCC.
Conforms to the purity criteria on food additives according to the current European Commission Regulation.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR