Hydrofluoric acid 40%, EMSURE® ISO for analysis, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

1.00338.0500 1.00338.1000 1.00338.2500 1.00338.9180
1.00338.0500EA 64.8 EUR
1.00338.0500 1.00338.1000 1.00338.2500 1.00338.9180
Hydrofluoric acid 40%, EMSURE® ISO for analysis, Supelco®
Hydrofluoric acid
Formula: HF
MW: 20.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8 (6.1) II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -44 °C
Boiling Point 112 °C
Density 1.13 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly acid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR