L(+)-Ascorbic acid

Supplier: Merck

Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum

MQ300

1.00468.0100 1.00468.0500 1.00468.1000
1.00468.0100EA 80.3 EUR
1.00468.0100 1.00468.0500 1.00468.1000
L(+)-Ascorbic acid
L(+)-Ascorbic acid
Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 553
Melting Pt: 190…192 °C
Density: 1.694 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
EINECS: 200-066-2
REACH: exempt
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 330 g/l (24 °C)
Melting Point 190 - 192 °C (decomposition)
Molar Mass 176.12 g/mol
Bulk Density 500 - 900 kg/m3
Vapor Pressure (20 °C)
Density 1.65 g/cm3 (20 °C)
pH 2.2 - 2.5 (50 g/l, H2O, 20 °C)
Ignition Point 380 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR