Orthophosphoric acid 85% 85, Suprapur® Trace_analysis, Supelco®

Supplier: Merck
Danger

Synonyms: Phosphoric acid

MQ100

1.00552.0250 1.00552.1000 1.00552.0500 1.00552.2500
1.00552.0250EA 258.82 EUR
1.00552.0250 1.00552.1000 1.00552.0500 1.00552.2500
Orthophosphoric acid 85% 85, Suprapur® Trace_analysis, Supelco®
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 21 °C
Boiling Point 158 °C
Vapor Pressure 2 hPa (20 °C)
Density 1.71 g/cm3 (20 °C)
pH  - 0.5 (100 g/l, H2O, 20 °C)
Viscosity kinematic 30.5 mm2/s (20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR