Salicylic acid 99.5-105.0% (by acidimetry on anhydrous substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Danger

1.00631.1000 1.00631.9024
1.00631.1000EA 521 EUR
1.00631.1000 1.00631.9024
Salicylic acid 99.5-105.0% (by acidimetry on anhydrous substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Salicylic acid
Formula: (HO)C₆H₄CO₂H
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 157…159 °C
Density: 1,443 g/cm³ (20 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 13,08411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1f695fa92656a135b5e1742f1cca28f4d255fb&vid=8f3aff6fb9a70e5dd194b8bc46019af74e48e90a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric; calculated on dried substance) 99.5 - 100.5 %
Assay (HPLC, calc.on dried substance) 98.0 - 102.0 %
Identity passes test
Appearance of solution (10 %; ethanol 96 %) passes test
Chloride (Cl) ≤ 0.0100 %
Sulfate (SO₄) ≤ 0.0200 %
Related substances / HPLC (EP) (4-Hydroxybenzoic acid) ≤ 0.1 %
Related substances / HPLC (EP) (4-Hydroxyisophthalic acid) ≤ 0.05 %
Related substances / HPLC (EP) (Phenol) ≤ 0.02 %
Related substances / HPLC (EP) (Largest unspecified impurity) ≤ 0.05 %
Related substances / HPLC (EP) (Sum of all impurities) ≤ 0.2 %
Related substances / HPLC (USP) (4-Hydroxybenzoic acid) ≤ 0.1 %
Related substances / HPLC (USP) (4-Hydroxyisophthalic acid) ≤ 0.05 %
Related substances / HPLC (USP) (Phenol) ≤ 0.02 %
Related substances / HPLC (USP) (Largest unspecified impurity) ≤ 0.05 %
Related substances / HPLC (USP) (Sum of all impurities) ≤ 0.2 %
Residual solvents (ICH Q3C) excluded by manufacturing process
Sulfated ash ≤ 0.05 %
Loss on drying ≤ 0.5 %
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
corresponds to Ph Eur, BP, USP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR