Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®

Supplier: Merck
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

M-Clarity™ quality level = MQ100

1.00658.2500 1.00658.1000
1.00658.2500EA 175 EUR
1.00658.2500 1.00658.1000
Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) =99.8%
Identity(IR) Conforms
Evaporation Residue = 3.0 mg/l
Water = 0.05%
Color =10 Hazen
GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)) = 3ng/ml
Suitability Residue Analysis

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR