n-Hexane ≥98%, SupraSolv® for GC-MS

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

1.00795.2500 1.00795.1000
1.00795.2500EA 202 EUR
1.00795.2500 1.00795.1000
n-Hexane ≥98%, SupraSolv® for GC-MS
n-Hexane
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94,3 °C
Density: 0,659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) (sum of hexane isomers plus methylcyclopentane) =98.0% (=99.8%)
Identity(IR) Conforms
Evaporation Residue = 3.0 mg/l
Water = 0.01%
Color =10 Hazen
GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)) = 3ng/ml
Suitability Residue Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR