Toluene 99,8, SupraSolv® GC_MS

Supplier: Merck
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

MQ100

1.00849.2500 1.00849.1000
1.00849.2500EA 154 EUR
1.00849.2500 1.00849.1000
Toluene 99,8, SupraSolv® GC_MS
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) =99.8%
Identity(IR) Conforms
Evaporation Residue = 3.0 mg/l
Water = 0.03%
Color =10 Hazen
GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)) = 3ng/ml
Suitability Residue Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR