Ethanol denatured, EMSURE® for analysis (denatured with 1% MEK), Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

1.00974.1011 1.00974.2511 1.00974.9025 1.00974.9180 1.00974.6025 1.00974.9950 1.00974.2500
1.00974.1011EA 46.2 EUR
1.00974.1011 1.00974.2511 1.00974.9025 1.00974.9180 1.00974.6025 1.00974.9950 1.00974.2500
Ethanol denatured, EMSURE® for analysis (denatured with 1% MEK), Supelco®
Ethanol denatured

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formula: H₃CCH₂OH
Density: 0,79 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Purity (GC) ≥ 99.5 %
Identity (IR) conforms
Free acid (as acetic acid) ≤ 0.001 %
Ethylmethylketone (GC) 0.8 - 1.3 %
Readily carbonizable substances conforms
Evaporation residue ≤ 0.001 %
Water ≤ 0.1 %
Al (Aluminium) ≤ 0.00005 %
B (Boron) ≤ 0.00005 %
Ba (Barium) ≤ 0.00001 %
Ca (Calcium) ≤ 0.00005 %
Cd (Cadmium) ≤ 0.000005 %
Co (Cobalt) ≤ 0.000002 %
Cr (Chromium) ≤ 0.000002 %
Cu (Copper) ≤ 0.000002 %
Fe (Iron) ≤ 0.00001 %
K (Potassium) ≤ 0.0001 %
Mg (Magnesium) ≤ 0.00001 %
Mn (Manganese) ≤ 0.000002 %
Ni (Nickel) ≤ 0.000002 %
Pb (Lead) ≤ 0.00001 %
Sn (Tin) ≤ 0.00001 %
Zn (Zinc) ≤ 0.00001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR