Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS
Danger

1.00986.1000 1.00986.2500 1.00986.6025 1.00986.6010 1.00986.9180 1.00986.1000DP 1.00986.1000_T 1.00986.2500_T 1.00986.6025_T 1.00986.6010_T 1.00986.9180_T 1.00986.9950
1.00986.1000EA 209 EUR
1.00986.1000 1.00986.2500 1.00986.6025 1.00986.6010 1.00986.9180 MERP1.00986.1000DP MERP1.00986.1000_T MERP1.00986.2500_T MERP1.00986.6025_T MERP1.00986.6010_T MERP1.00986.9180_T 1.00986.9950
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Ethanol

For full details of specification and hazards please see www.vwr.com


Other residual solvents (ICH Q3C) excluded by production process.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities).
Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Certifications: Conforms to Ph Eur, BP, JP, USP

Formula: C₂H₅OH
Boiling Pt: 78,3 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0,7895 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specification Test Results

Assay (m/m) ≥ 99.2 %
Assay (V/V) ≥ 99.5 %
Identity (IR) conforms
Identity 1 (ChP) conforms
Appearance conforms
Clarity of solution conforms
Color of solution conforms
Clarity and color of solution (ChP) conforms
Acidity or alkalinity ≤ 30 ppm
Acidity or alkalinity (ChP) conforms
Density (d 20 °C/20 °C) 0.790 - 0.793
Density (d 15,56 °C/15,56 °C) ≤ 0.7962
Density (d 15 °C/15 °C) 0.794 - 0.797
Specific gravity (d 15 °C/15 °C) 0.79422 - 0.79679
Absorbance (at 240 nm) ≤ 0.40
Absorbance (between 250 nm and 260 nm) ≤ 0.30
Absorbance (between 270 nm and 340 nm) ≤ 0.10
The spectrum shows a steadily descending curve with no observable peaks or shoulders conforms
Absorbance (ChP) (at 240 nm) ≤ 0.08
Absorbance (ChP) (between 250 nm and 260 nm) ≤ 0.06
Absorbance (ChP) (between 270 nm and 340 nm) ≤ 0.02
Volatile impurities (GC) (Acetaldehyde and Acetal) ≤ 10 ppm
Volatile impurities (GC) (Benzene) ≤ 2 ppm
Volatile impurities (GC) (Methanol) ≤ 100 ppm
Volatile impurities (GC) (Total of other impurities) ≤ 300 ppm
Cyclohexane (GC) ≤ 50 ppm
Evaporation residue ≤ 25 mg/l
Non-volatile matter ≤ 25 mg/l
Bacterial endotoxins ≤ 2.5 I.U./ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR