L-Alanine, Millipore®

Supplier: Merck

Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine

M-Clarity™ quality level = MQ100

1.01007.0025 1.01007.0100 1.01007.9010 1.01007.1000
1.01007.0025EA 58.3 EUR
1.01007.0025 1.01007.0100 1.01007.9010 1.01007.1000
L-Alanine, Millipore®
L-Alanine

Soluble in water and mineral acids; insoluble in organic solvents.

Formula: C₃H₇NO₂
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 79 °C
Density: 1,4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064410
CAS Number: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR