Alizarin complexone dihydrate Fluoride_determination, Supelco®

Supplier: Merck

Synonyms: 3-Bis(carboxymethyl)aminomethyl-1,2-dihydroxyanthraquinone dihydrate, Alizarin fluorine blue dihydrate, 3,4-dihydroxyanthraquinon-2-ylmethyliminodi(acetic acid) dihydrate, 2-[(3,4-dihydroxy-9,10-dioxoanthracen-2-yl)methyl-(2-oxido-2-oxoethyl)azaniumyl]acetate dihydrate, Alizarine-3-methylamine-N,N-diacetic acid dihydrate

MQ100

1.01010.0005
1.01010.0005EA 384 EUR
1.01010.0005
Alizarin complexone dihydrate Fluoride_determination, Supelco®
Alizarin complexone dihydrate
Formula: C₁₉H₁₅NO₈·2H₂O
MW: 421.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149067
CAS Number: 455303-00-1
EINECS: 223-544-2

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Molar Mass 421.36 g/mol
Bulk Density 540 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR