Aluminium potassium sulphate dodecahydrate, EMSURE® ACS, Reag. Ph. Eur. analysis_for, Supelco®

Supplier: Merck

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

MQ300

1.01047.1000 1.01047.9025
1.01047.1000EA 88.3 EUR
1.01047.1000 1.01047.9025
Aluminium potassium sulphate dodecahydrate, EMSURE® ACS, Reag. Ph. Eur. analysis_for, Supelco®
Aluminium potassium sulphate dodecahydrate
Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92.5
Density: 1.72 g/cm³ (20 °C)
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now

Specification Test Results

Solubility 139 g/l (20 °C)
Melting Point 92.5 °C
Molar Mass 474.39 g/mol
Bulk Density 900 kg/m3
Density 1.75 g/cm3 (20 °C)
pH 3.0 - 3.5 (100 g/l, H2O, 20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR