Ammonium peroxodisulphate (APS), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

1.01201.0500 1.01201.1000 1.01201.5000 1.01201.9050 1.01201.9012
1.01201.0500EA 76.8 EUR
1.01201.0500 1.01201.1000 1.01201.5000 1.01201.9050 1.01201.9012
Ammonium peroxodisulphate (APS), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Ammonium peroxodisulphate (APS)
Formula: (NH₄)₂S₂O₈
Melting Pt: 120 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 620 g/l (20 °C)
Melting Point 120 °C (decomposition)
Molar Mass 228.19 g/mol
Bulk Density 900 kg/m3
Density 1.98 g/cm3 (20 °C)
pH 3.2 (100 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR