Absorbent granules, Chemizorb® granules
Supplier: Merck
1.01568.1000 1.01568.5000 1.01568.9021
1.01568.1000EA 45.38 EUR
1.01568.1000 1.01568.5000 1.01568.9021
Absorbent granules, Chemizorb® granules
Spill Control Materials
Chemizorb granules is a coarse grained material based on chemically-inert porous minerals or synthetic co-polymers (grain size over 1 mm (95%), bulk density of 400 kg/m³).
  • Able to absorb up to 100% of its own weight
  • Low viscosity liquids are absorbed instantaneously, highly viscous media (glycerine, oils) within a few minutes
  • Good dosing is possible thanks to the coarse grained format
  • Granule form is used in all those places where it is not suitable to use the powder form, for example, in draughty rooms or outside
It is insoluble in water and in allMore Product Information
Limited Time Sale
Select items below are currently on sale
Order Now
SPECIFICATIONS
Environmentally Preferable