Barium carbonate, Technipur®

Supplier: Merck
Warning

1.01711.1000 1.01711.9050
1.01711.1000EA 85.3 EUR
1.01711.1000 1.01711.9050
Barium carbonate, Technipur®
Barium carbonate
Formula: BaCO₃
Melting Pt: 1350 °C
Density: 3,89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 13,00972

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR