Barium hydroxide octahydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

1.01737.0500 1.01737.9050
1.01737.0500EA 136 EUR
1.01737.0500 1.01737.9050
Barium hydroxide octahydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Barium hydroxide octahydrate
Formula: Ba(OH)₂·8H₂O
Boiling Pt: 780 °C (1013 hPa)
Melting Pt: 78 °C
Density: 2,18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 2923
ADR: 8 (6.1),II
Merck Index: 13,00980

Order Now

Specification Test Results

Solubility 72 g/l (20 °C)
Melting Point 78 °C
Molar Mass 315.48 g/mol
Bulk Density 900 - 1100 kg/m3
Density 2.18 g/cm3 (20 °C)
pH 12.5 (50 g/l, H2O, 20 °C) (slurry)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR