Bromothymol blue sodium salt, water soluble indicator, Supelco®

Supplier: Merck

Synonyms: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate

M-Clarity™ quality level = MQ200

1.01895.0010
1.01895.0010EA 124 EUR
1.01895.0010
Bromothymol blue sodium salt, water soluble indicator, Supelco®
Bromothymol blue sodium salt
Formula: C₂₇H₂₇Br₂NaO₅S
Melting Pt: > 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00077263
CAS Number: 34722-90-2
EINECS: 252-169-7

Order Now

Specification Test Results

Molar Mass 646.36 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR