1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate, Millipore®

Supplier: Merck
Warning

Synonyms: Catechol-3,5-disulphonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt monohydrate, Tiron monohydrate, 3,5-Pyrocatecholdisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulphonate monohydrate

MQ200

1.01922.0100
1.01922.0100EA 619 EUR
1.01922.0100
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate, Millipore®
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate
Formula: C₆H₄Na₂O₈S₂·1H₂O
MW: 332.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149531
CAS Number: 149-45-1
EINECS: 205-741-5

Order Now

Specification Test Results

Solubility 1350 g/l (20 °C)
Melting Point 300 °C
Molar Mass 332.22 g/mol
Bulk Density 465 kg/m3
pH 5.0 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR