Silica gel 1 - 4 mm Drying_agent, with moisture indicator_brown gel, Supelco®

Supplier: Merck


MQ200

1.01972.9025 1.01972.5000 1.01972.1000
1.01972.9025EA 0 EUR
1.01972.9025 1.01972.5000 1.01972.1000
Silica gel 1 - 4 mm Drying_agent, with moisture indicator_brown gel, Supelco®
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR