Zinc, Certipur® Reag. Ph. Eur. volumetric standard, secondary reference material, for complexometry traceable to NIST, Supelco®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

1.02409.0100 1.02409.0103
1.02409.0100EA 203 EUR
1.02409.0100 1.02409.0103
Zinc, Certipur® Reag. Ph. Eur. volumetric standard, secondary reference material, for complexometry traceable to NIST, Supelco®
Zinc

Merck have introduced into the range of Certipur® standards some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: Zn
Boiling Pt: 906 °C (1013 hPa)
Melting Pt: 419 °C
Density: 7,14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9,III
Merck Index: 12,10255

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 419 °C
Molar Mass 65.39 g/mol
Boiling Point 906 °C (1013 hPa)
Density 7.13 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR