Copper(I) chloride, EMSURE® ACS for analysis, Supelco®

Supplier: Merck
Danger

Synonyms: Cuprous chloride, Copper chloride

M-Clarity™ quality level = MQ300

1.02739.0250 1.02739.9025
1.02739.0250EA 852 EUR
1.02739.0250 1.02739.9025
Copper(I) chloride, EMSURE® ACS for analysis, Supelco®
Copper (I) chloride
Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1367
Melting Pt: 430
Density: 4.145 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8 III
Merck Index: 13,02690

Order Now

Specification Test Results

Solubility 0.001 - 0.1 g/l
Melting Point 422 °C
Molar Mass 99 g/mol
Bulk Density 1600 - 1800 kg/m3
Boiling Point 1367 °C
Density 4.14 g/cm3 (25 °C)
pH 5 (50 g/l, H2O, 20 °C) (slurry)
Hazard Class 8
Hazard Class Description Corrosive Material

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR