Bromophenol red, Millipore®

Supplier: Merck


M-Clarity™ quality level = MQ200

1.03023.0005
1.03023.0005EA 290 EUR
1.03023.0005
Bromophenol red, Millipore®
Bromophenol red
Formula: C₁₉H₁₂Br₂O₅S
Melting Pt: 230 °C
MDL Number: MFCD00023014
CAS Number: 2800-80-8
EINECS: 220-538-1

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 230 °C (decomposition)
Molar Mass 512.18 g/mol
Bulk Density 770 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR